Skip to Content
MilliporeSigma

QGARDTL804

Q-Gard® 8 Polishing Cartridge

Removes trace ions to produce high-resistivity water. For Milli-Q® CLX 8Series systems. This product replaces QGARDTXL04.

Synonym(s):

Q-Gard® 8 Purification Cartridge

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.85
UNSPSC Code:
41104212
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polypropylene and polyethylene cartridge, resin (Virgin ion-exchange resins (for deionization))

packaging

pkg of 1 unit

manufacturer/tradename

Q-Gard®

parameter

at 10-40 °C (air), at 5-35 °C (water)

H × width × D

52.8 cm (20.8 in.) × 15.5 cm (6.1 in.) × 7.9 cm (3.1 in.)

capacity

32 g/L ion exchange capacity(Dynamic ion exchnage capacity

NaCl), 43 g/L total exchange capacity(NaCl

NaCl)

fitting

#11 female inlet and outlet for #11 (O-rings)

compatibility

for use with Milli-Q® CLX (8 series)

shipped in

ambient

General description

Q-Gard® XL Polishing Cartridge contains a combination of purification media carefully selected to remove trace ions to produce high-resistivity water.
The cartridge is constructed with clean and high-purity materials to minimize leaching of extractable contaminants.
For use with Milli-Q® CLX 8Series systems.

Application

This polishing cartridge contains virgin ion-exchange resins to remove trace ions. It is recommended for any laboratory work requiring very low levels of ions from high volume systems.

Features and Benefits

  • Q-Gard® XL Polishing Cartridge is tailored to the feed water source to optimize the performance of downstream purification media to ensure consistent results
  • Easy to replace - every 6 month is recommended for optimal results
  • e-Sure tag RFID technology for automatic recognition of the right pack. Automatically fills and archives pack parameters for full traceability
  • Genuine consumable manufactured in our ISO 9001-certified facility
  • Quality certificate available online (available in the bottom part of this web page in the "Documentation" section)

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library