MilliporeSigma
ZCLXL8T0C
TOC monitor upgrade kit for Milli-Q® CLX 8Series systems
Enables measurements of TOC in loop water. For Milli-Q® CLX 8Series water purification systems. This product replaces ZAFSL0T0C.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
SB.51
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® CLX (8 series)
General description
Installed within the purification system, the in-line TOC (total organic carbon) monitor verifies TOC levels of system water is less than 500 ppb.
Features and Benefits
  • TOC monitor saves time and money and avoids the need for duplicate assays or repeated instrument calibrations.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)