Skip to Content
MilliporeSigma

ZLXLINDRES

Emergency bypass resistivity indicator kit

For use with Milli-Q® CLX 8Series water purification systems. This product replaces ZSNAAFSQUAL.

Synonym(s):

water purification accessory

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104206
eCl@ss:
32160703
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Milli-Q® CLX

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® CLX (8 series)

General description

The emergency bypass resistivity indicator kit is used during an emergency bypass procedure to check water quality in the loop and indicates when to change the Q-Gard® cartridge to maintain water quality.

Features and Benefits

  • Easy to use if emergency bypass is needed
  • Measures the level of resistivity in the loop. Green indicates the level is good and Red if it′s not good.
  • The Q-Gard® Purification Cartridge must be changed when the indicator′s light turns red

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library