Skip to Content
MilliporeSigma

PBKH9AA72B0E

PharmaBlock (USA), Inc.

cis-2-methoxycarbonylcyclopropanecarboxylic acid

Synonym(s):

(1S,2R)-2-(methoxycarbonyl)cyclopropanecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O4
CAS Number:
Molecular Weight:
144.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8O4/c1-10-6(9)4-2-3(4)5(7)8/h3-4H,2H2,1H3,(H,7,8)/t3-,4+/m0/s1

Inchi Key

IRRAJLZEAOBJIV-IUYQGCFVSA-N

Purity

97%

Storage temp.

Refrigerated storage

Country of Origin

CN

Physical Form

Solid




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library