Skip to Content
MilliporeSigma

RIE156317721

Rieke Metals, Inc.

2,9-dichloro-1-nonene

Synonym(s):

2,9-dichloro-1-nonene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
195.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H16Cl2/c1-9(11)7-5-3-2-4-6-8-10/h1-8H2

Inchi Key

YOSLDMVBPIVEPQ-UHFFFAOYSA-N

Purity

97%

Country of Origin

US



Still not finding the right product?

Explore all of our products under 2,9-dichloro-1-nonene




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library