MilliporeSigma
All Photos(1)

14373C

SAFC

Stemline® XF MSC Medium Kit

Serum-free, liquid, sterile-filtered, suitable for cell culture

Synonym(s):
Mesenchymal Stem / Stromal Cell Medium

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable (suspension)

shipped in

wet ice

storage temp.

2-8°C

General description

Developed to promote the expansion of human mesenchymal stem/stromal cells (MSCs) in both planar and suspension culture formats, Stemline XF MSC Medium demonstrates better cell expansion performance than both α-MEM supplemented with human platelet lysate and commercially available products when culturing MSCs on microcarriers. Stemline XF MSC Medium is compatible with MSCs derived from multiple tissues and has been performance verified in 3 L stirred tank bioreactors. Additionally, both characterization and functional assays (flow cytometry, IDO assay, and differentiation to adipocytes, chondrocytes, and osteocytes) confirm that Stemline XF MSC Medium maintains MSCs in the appropriate multipotent state.

Stemline XF MSC Medium Kit consists of one (1) 500 mL bottle of Stemline XF MSC Basal Medium (14371C) and one (1) 5 mL vial of Stemline XF MSC Supplement (14372C). The Basal Medium and Supplement may also be purchased separately in the same sizes or a larger 5 L bag (Basal Medium) and 50 mL bottle (Supplement).Stemline XF MSC Medium has been designed with your long-term needs for clinical manufacturing in mind, with a sustainable raw material supply chain, custom packaging options, and careful material and manufacturing process selection. The product is xeno-free and produced in an ISO9001-certified facility.

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
Stemline is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service