Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62353

Millipore

Listeria Enrichment Broth according to FDA/IDF-FIL

NutriSelect® Plus, suitable for microbiology

Synonym(s):

LE Broth, TSYEB, Tryptic Soy Yeast Extract Broth

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171607
NACRES:
NA.85

agency

according to GB 4789.30-2016

Quality Level

description

for the selective enrichment of Listeria.

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein peptone, 17 g/L
dipotassium hydrogen phosphate, 2.5 g/L
D(+)-glucose, 2.5 g/L
sodium chloride, 5 g/L
soy peptone, 3 g/L
yeast extract, 6 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages
sample preparation

microbiology

suitability

selective for Listeria spp.

Application

for the selective enrichment of Listeria

Preparation Note

Dissolve 18 g in 500 ml distilled water. Autoclave at 121°C for 15 minutes. Dissolve the contents of a vial of Listeria selective supplement (Cat. No. 62348 or 1.11781.0010) in 1 ml sterile distilled water and add to the broth cooled to below 50°C. Mix carefully by gentle swirling.

Other Notes

Listeria monocytogenes in raw milk: detection, incidence and pathogenicity

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lovette, J., et al.
Journal of Food Protection, 50, 188-188 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service