About This Item
biological source
synthetic (organic)
Quality Level
agency
USP/NF
meets USP testing specifications (Not intended for use in preparing injectable dosage forms.)
assay
98.0-102.0% anhydrous basis
98.0-102.0%
form
powder
impurities
<0.002% heavy metal
<3 ppm Arsenic
color
white
application(s)
pharmaceutical (small molecule)
storage temp.
room temp
SMILES string
OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
InChI
1S/2C6H12O7.Ca/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;/h2*2-5,7-11H,1H2,(H,12,13);/q;;+2/p-2/t2*2-,3-,4+,5-;/m11./s1
InChI key
NEEHYRZPVYRGPP-IYEMJOQQSA-L
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service