Skip to Content
MilliporeSigma

HIPC-1

High purity carbon certified reference material for mass fraction of carbon, carbon isotope delta and elemental impurities

certified reference material

Synonym(s):

High purity carbon certified reference material for mass fraction of carbon, carbon isotope delta and elemental impurities

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

traceable to (National Research Council of Canada (NRC))

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

neat

storage temp.

room temp

General description

NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.
HIPC-1 is a high purity carbon Certified Reference Material (CRM). A unit of HIPC-1 consists of approximately 0.25 g of high purity carbon rod. This material is intended as a primary standard for the determination of the mass fraction of carbon, and for carbon isotope delta measurements. For more information, please refer to the Certificate of Analysis or Safety Data Sheet.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library