MilliporeSigma
All Photos(1)

Documents

MIC003

Supelco

E. coli in Water PT- Quantitative WP

Proficiency Testing Material

NACRES:
NA.85

Quality Level

packaging

set of 2 ea

matrix

Vitroid

application(s)

environmental
food and beverages

shipped in

wet ice

storage temp.

−20°C

suitability

Escherichia coli
coliforms

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
MIC007MIC011MIC005
application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

suitability

Escherichia coli, coliforms

suitability

Escherichia coli, coliforms

suitability

Enterococcus spp., Streptococcus spp.

suitability

Enterococcus spp., Streptococcus spp.

packaging

set of 2 ea

packaging

set of 2 ea

packaging

set of 2 ea

packaging

set of 2 ea

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

E. coli in Water PT - Quantitative WP is produced in accordance with ISO/IEC 17043:2010. This sample is manufactured according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WP program is designed to meet the quality assurance requirements in compliance with Discharge Monitoring Report - Quality Assurance Program (DMR-QA), thereby ensuring the analytical competence of your laboratory.

Type of PT Scheme: Water analysis - Water Pollution (WP)

Matrix: Water Pollution/Wastewater

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of E. coli in Water PT- Quantitative WP proficiency testing standard for E. coli testing in water can be found in the reporting packet which will be available to download at PT Portal.

Application

E. coli in Water PT - Quantitative WP proficiency testing material is designed for the quantitative determination of E. coli in wastewater samples.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Packaging

Set contains 2 sterile screw cap vials each containing a Vitroid (a small disk containing the possible organisms) and 2 - 99mL bottles of phosphate buffer.
(Please note, buffer solution may not be provided with your sample. If this causes any concern, please contact PT Service ptservice@milliporesigma.com.)

Preparation Note

Description
  • This PT sample is designed for the quantitative analysis of total and/or fecal coliform and E. coli.
  • The PT sample is provided in duplicate as a stable dehydrated disc requiring hydration. The additional sample is provided for mishaps in the lab. TNI prohibits the practice of multiple tests on a PT sample unless it is normal laboratory practice and multiple analysis is conducted on normal day to day samples.
  • If you are following IDEXX methods for Quantitray follow the directions provided with the media (suggests using Type 1 DI water.)
  • When prepared according to instructions, the concentration range of the sample will be between 20-2400 CFU/100mL as per TNI Fields of Testing, January 4, 2010.
  • The dehydrated disc sample is stable for a time exceeding the study schedule.General Information

General Information
  • Due to the sensitivity of microorganisms and short holding times after hydration, please read entire instructions prior to preparation.
  • This sample is a single disc and the possible concentration range is quite broad, it may be necessary to analyze the sample in a dilution series using a general phosphate buffer solution.
  • This sample may be analyzed by any method appropriate for coliforms.

Sample Preparation
  • Prepare your lavatory standard buffer or saline solution, this is your hydration fluid (see note below).
  • Allow the capped sample vial to equilibrate to room temperature.
  • Open the sample vial and aseptically transfer the disc to 100 ml of the hydrating fluid. NOTE: The sample is the colored disc within the cryotube. It will either be on top of cotton with opaque desiccant beads below the cotton or free moving in a cryotube that contains a desiccant tablet within the cap. The opaque desiccant beads and desiccant tablet are to keep the container dry and are not biologicals.
  • Allow the disc to dissolve completely this may take up to 15 minutes. Mix by inverting gently several times, shaking may cause the cap to leak.
  • This represents the sample for analysis. Incubate at the temperature required by your method. Temperature requirements are rigorous, higher temperatures will kill the organisms and lower temperatures may suppress growth. Minimum incubation time is 24 hours (or 18 hours dependent on method and media.) Further incubation time may be necessary depending on the method and media. Further dilutions are recommended to achieve a proper count, but base all counts on the whole volume sample. If you have questions, please contact ptservice@milliporesigma.com.
Note: We are no longer supplying Phosphate buffer for Micro samples. Please proceed to use your laboratory′s standard buffer or saline solution. If your laboratory does not have buffer in-house, we recommend using P3813, Phosphate Powder which can be purchased at SigmaAldrich.com.

Storage and Stability

Please refer to the sample label for storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

This PT sample is designed for the quantitative analysis of total and/or fecal coliform and E. coli. The PT sample is provided in duplicate as a stable dehydrated vitroid requiring hydration. The additional sample is provided for mishaps in the lab. Store the PT sample as directed. The dehydrated vitroid™ sample is stable for a time exceeding the study schedule.

Proficiency Testing Schedule (2022-2023)

Wastewater
WP22-1 January 12 – February 25, 2022
WP22-2 March 23 - May 6, 2022
WP22-3 July 6 - August 19, 2022
WP22-4 September 21 - November 4, 2022

WP23-1 January 11 – February 24, 2023
WP23-2 March 22 – May 5, 2023
WP23-3 July 5 – August 18, 2023
WP23-4 September 20 – November 3, 2023


Quick-Turn Study (on demand proficiency test that is offered depending on lot availability)

DMR-QA (US customers only)
DMR-QA 43 2023 Dates TBD

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Total coliforms 20.0 - 2400 CFU/100mL
    Total Coliform, MPN 20.0 - 2400 MPN/100 mL
    Escherichia coli 20.0 - 2400 CFU/100mL
    Escherichia coli, MPN 20.0 - 2400 MPN/100 mL
    Fecal coliforms 20.0 - 2400 CFU/100mL
    Fecal coliform, MPN 20.0 - 2400 MPN/100 mL

related product

Product No.
Description
Pricing

Storage Class Code

6.2 - Infectious substances

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 1

1 of 1

Supelco

Supelco

PE3050

Residue - WP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service