Skip to Content
MilliporeSigma

NIST2193B

Calcium Carbonate pH Standard (used as saturated calcium hydroxide solution) (pH Standard)

NIST®SRM®

Synonym(s):

Calcium carbonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein/REAXYS Number:
8008338
MDL number:
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 30 g

manufacturer/tradename

NIST®

density

2.93 g/mL at 25 °C (lit.)

application(s)

general analytical

format

neat

storage temp.

15-25°C

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology


Storage Class

13 - Non Combustible Solids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library