MilliporeSigma
All Photos(1)

Documents

PE1130

Supelco

Demand - WP

Proficiency Testing Material, Concentrate for dilution to 1 L

NACRES:
NA.24

Quality Level

packaging

ampule of 2 × 20 mL

concentration

in water

application(s)

environmental

format

matrix material

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PE3130PE3050PE1153
Supelco

Supelco

PE1130

Demand - WP

Supelco

Supelco

PE3130

Demand - Whole Volume - WP

Supelco

Supelco

PE3050

Residue - WP

Supelco

Supelco

PE1153

Nitrite - WP

format

matrix material

format

matrix material

format

matrix material

format

matrix material

packaging

ampule of 2 × 20 mL

packaging

bottle of 1 × 500 mL

packaging

bottle of 1 × 500 mL

packaging

ampule of 2 × 2 mL

concentration

in water

concentration

in water

concentration

in water

concentration

in water

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

General description

Demand - WP proficiency testing material is produced in accordance with ISO/IEC 17043:2010. Also referred to as non-Potable water or Water Pollution sample, this product is produced according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The higher criteria from the TNI tables mimic the levels common to wastewater samples. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. Our WP program is designed to meet the quality assurance requirements in compliance with Discharge Monitoring Report - Quality Assurance Program (DMR-QA), thereby ensuring the analytical competence of your laboratory.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Water analysis - Water Pollution (WP)

Matrix: Water Pollution/Wastewater

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Demand - WP can be found in the reporting packet which will be available to download at PT Portal.

Application

Demand - WP is a non-potable water (NPW) PT standard, suitable for testing analytes in water pollution/wastewater.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • This PT sample is packaged as a 20 mL concentrate.
  • The solvent for this sample is water.
  • The PT sample has been autoclaved for preservation.

Sample Preparation

  • To prepare the solution, partially fill a 1 L volumetric flask with reagent-grade water.
  • Open ampule PE1130-20ML (be careful to avoid cuts on glass) and transfer 10 mL from the ampule into the flask.
  • Fill the volumetric flask to volume with reagent-grade water and mix well.
  • The sample is now ready for immediate analysis.
  • Analyze the solution to determine BOD or CBOD first. If COD and TOC cannot be determined within two hours after sample preparation, an aliquot should be preserved (pH<2) using 0.2 mL of concentrated sulfuric acid per 100 mL. This aliquot should be stored at 4 °C and analyzed as soon as possible within the 28-day maximum holding time.

NOTES: For proper oxygen depletion of BOD and CBOD, the sample should be tested in a dilution series. Use natural surface water or diluted domestic sewage for seeded dilution water and avoid entrainment of air. For analysis of CBOD, the use of a nitrifying inhibitor is recommended. Be sure to determine the "blank" BOD of your seeded dilution water so that the proper seed correction can be made.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency Testing Schedule (2022-2023)

Wastewater
WP22-1 January 12 – February 25, 2022
WP22-2 March 23 - May 6, 2022
WP22-3 July 6 - August 19, 2022
WP22-4 September 21 - November 4, 2022

WP23-1 January 11 – February 24, 2023
WP23-2 March 22 – May 5, 2023
WP23-3 July 5 – August 18, 2023
WP23-4 September 20 – November 3, 2023

DMR-QA (US customers only)
DMR-QA 43 2023 Dates TBD

Quick-Turn Study (on demand proficiency test that is offereddepending on lot availability)

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    5-Day BOD 18-230 mg/L
    Carbonaceous BOD (CBOD) 18-230 mg/L
    Chemical oxygen demand (COD) 30-250 mg/L
    Dissolved organic carbon (DOC) 6-100 mg/L
    Total organic carbon (TOC) 6-100 mg/L

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 10

1 of 10

Supelco

Supelco

PE1041

Minerals - WP

Supelco

Supelco

QC1388

Demand - WS

Supelco

Supelco

PE1077

Dissolved Oxygen - WP

Supelco

Supelco

PE1132

Trace Metals 1 - WP

Supelco

Supelco

QC3130

Demand - Whole Volume

Supelco

Supelco

PE1083

Oil and Grease - WP

Supelco

Supelco

QC1051

Complex Nutrients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service