MilliporeSigma
All Photos(1)

Documents

PE1207

Supelco

Fatty Acid Methyl Esters (FAME)

Proficiency testing material

Quality Level

application(s)

food and beverages

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PE120049454-USPE016TIS
Supelco

Supelco

PE1200

Aflatoxin Mixture

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

General description

Fatty Acid Methyl Esters (FAME) is produced in accordance with ISO/IEC 17043:2010. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Most water PT samples are available as concentrates or ready-to-use volumes giving the end-user flexibility in selecting samples that will fit their exact needs. This PT portfolio is specially designed to evaluate the quality and demonstrate the reliability of the data for measuring the food quality, improving the customer satisfaction via adhering by the government regulations.

Matrix: Food Dichloromethane

All information regarding the use of Fatty Acid Methyl Esters (FAME) proficiency testing standard can be found in the reporting packet which will be available to download at PT Portal.

Application

Fatty Acid Methyl Esters (FAME) as a proficiency testing material is used to test fatty acid methyl esters in food samples.

Have any questions? Explore our FAQs
For more information on this product, please contact our PT Service Team.

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • This sample contains 37 fatty acid methyl esters in dichloromethane. No derivatization is required for this sample.
  • The sample contains 1.2 mL of sample.
  • The sample is designed for GC analysis.

Sample preparation

  • The sample needs no special treatment before use.
  • Use your standard sample preparation procedure.

Analysis

  • Use your standard analysis method to analyze the sample.
  • Report your results as ug/mL in the Proficiency Testing Portal.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency Testing Schedule (2021-2022)

Food
FOOD22-3 October 19 - December 02, 2022

FOOD23-3 July 19 – September 1, 2023

Quick-Turn Study Not Available for this Product

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Myristoleic Acid Methyl Ester (Methyl Myristoleate) 20-150 μg/μL
    cis-10-Pentadecenoic Acid Methyl Ester 20-150 μg/μL
    Linolelaidic Acid Methyl Ester (Methyl Linolelaidate) 20-150 μg/μL
    Gamma-Linolenic Acid Methyl Ester (Methyl ?-Linolenate) 20-150 μg/μL
    Methyl cis-5,8,11,14-Eicosatetraenoic ( Methyl Arachidonate) 20-150 μg/μL
    All cis-4,7,10,13,16,19-Docosahexaenoate 20-150 μg/μL
    Methyl cis-5,8,11,14,17-Eicosapentaenoate 20-150 μg/μL
    Methyl Butyrate 40-150 μg/μL
    Methyl Hexanoate 40-150 μg/μL
    Methyl Octanoate 40-150 μg/μL
    Methyl Decanoate (Caprate) 40-150 μg/μL
    Methyl Undecanoate 20-150 μg/μL
    Methyl Laurate 40-150 μg/μL
    Methyl Tridecanoate 20-150 μg/μL
    Methyl Myristate 40-150 μg/μL
    Methyl Pentadecanoate 20-150 μg/μL
    Methyl Palmitate 60-150 μg/μL
    Methyl Heptadecanoate 20-150 μg/μL
    Methyl Stearate 40-150 μg/μL
    Methyl Heneicosanoate 20-150 μg/μL
    Methyl Behenate 40-150 μg/μL
    Methyl Tricosanoate 20-150 μg/μL
    Methyl Palmitoleate (Methyl cis 9-Hexadecenoate) 20-150 μg/μL
    cis-9-Oleic Acid Methyl Ester (Methyl Oleate) 40-150 μg/μL
    Methyl Linoleate 20-150 μg/μL
    Methyl Erucate (cis-13-Docosenoate) (Methyl cis-13-Docosenoate) 20-150 μg/μL
    Methyl Nervonate (Methyl cis-15-Tetracosenoate) 20-150 μg/μL
    Methyl Arachidate 40-150 μg/μL
    Methyl Lignocerate (Methyl Tetracosanoate) 40-150 μg/μL
    Trans-9-Elaidic Acid Methyl Ester (Methyl Elaidate) 20-150 μg/μL
    Methyl Linolenate 20-150 μg/μL
    Methyl cis-11 Eicosenoate 20-150 μg/μL
    cis-10-Heptadecenoic Acid Methyl Ester (Methyl cis-10-Heptadecenoate) 20-150 μg/μL
    cis-11,14-Eicosadienoic Acid Methyl Ester 20-150 μg/μL
    cis-11,14,17-Eicosatrienoic Acid Methyl Ester 20-150 μg/μL
    cis-8,11,14-Eicosatrienoic Acid Methyl Ester 20-150 μg/μL
    cis-13,16-Docosadienoic Acid Methyl Ester 20-150 μg/μL
    See All (37)

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service