MilliporeSigma
All Photos(1)

Documents

PE1364

Supelco

Anions - WS

Proficiency Testing Material, (Concentrate for dilution to 500 mL)

Synonym(s):
WS - Anions
NACRES:
NA.24

Quality Level

packaging

ampule of 2 × 20 mL

concentration

in water

application(s)

environmental

format

matrix material

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PE3364PE3060PE1541
Supelco

Supelco

PE1364

Anions - WS

Supelco

Supelco

PE3364

Anions - Whole Volume - WS

Supelco

Supelco

PE3060

Anions - Whole Volume - WP

Supelco

Supelco

PE1541

Sulfite in Water - WS

format

matrix material

format

matrix material

format

matrix material

format

matrix material

packaging

ampule of 2 × 20 mL

packaging

bottle of 1 × 500 mL

packaging

bottle of 1 × 500 mL

packaging

ampule of 2 × 20 mL

concentration

in water

concentration

in water

concentration

in water

concentration

in water

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

General description

Anions - WS proficiency testing material is produced in accordance with ISO/IEC 17043:2010. Also referred to as Potable Water or Water Supply sample, this sample is produced according to TNI requirements. However, please note, this sample may include non-TNI analytes as well. The lower criteria from the TNI tables mimic the levels common to drinking water samples. The PT portfolio allows our test components to offer significant performance data to monitor and prove competency to customers, accreditation bodies or both.

Supelco® is a global proficiency testing (PT) provider that manufactures best-in-class environmental and pharmaceutical PT products. Our WS program is designed to meet the quality assurance requirements in compliance with Safe Drinking Water Act (SDWA), thereby ensuring the analytical competence of your laboratory or drinking water facility.
These samples are typically provided in duplicate for use in case of lab mishap, for QC purposes or to analyze multiple methods.

Type of PT Scheme: Water analysis - Water Supply (WS)

Matrix: Water Supply/Drinking water

Analyte/s: Values of analytes vary lot to lot.

All information regarding the use of Anions - WS Proficiency Testing standard can be found in the reporting packet which will be available to download at PT Portal.

Application

Anions - WS proficiency testing material is suitable for testing the anions in water supply/drinking water.

Have any questions? Explore our FAQs.
For more information on this product, please contact our PT Service Team..

Features and Benefits

We have a vast portfolio of environmental PT schemes with "fit-for-purpose" samples to help you meet the needs of the laboratory.

Our proficiency testing (PT) program is a benchmark to:

  • provide staff with an insight into their laboratory′s performance, to verify the accuracy and reliability of their testing process
  • provide a comparison of performance with that of other laboratories, nationally and internationally
  • test and educate staff, provide a better understanding of the impact of incorrect results and identify areas where there may be problems
  • provide an independent evaluation of laboratory performance
  • demonstrate to clients, colleagues and accreditation bodies that there is a commitment to quality

Preparation Note

Description

  • This PT sample is packaged as a 20 mL concentrate.
  • The solvent for this sample is water.
  • The PT sample has been autoclaved for preservation.
  • The sample must be stored at 4 °C.

Sample preparation

  • All constituents are present in soluble form.
  • Partially fill a 500 mL volumetric flask with reagent-grade water (near 20 ºC).
  • Open ampule PE1364-20ML (be careful to avoid cuts on glass) and transfer 10.0 mL of concentrate to the volumetric flask.
  • Fill the flask to volume with reagent-grade water and mix well.
  • The sample is now ready for immediate analysis.
  • Sealed ampules were autoclaved to preserve the solutions inside.
  • This preservation treatment is not effective after the ampule is opened. Therefore, the sample should be analyzed as soon as possible after sample preparation following the preservation and holding times as specified in the method.

Storage and Stability

Please refer to sample label for expiration date, specific storage temperature, and specific storage conditions.

Other Notes

Are you new to Supelco® Proficiency Testing? Be sure to register your account details in the PT Portal before placing your order.
This will provide access to data entry, reporting packets and final reports. Registrations will be approved shortly after your order is placed. For further questions, write to us at ptservice@milliporesigma.com.

When placing your order online, please indicate your study preference under the ′your reference′ column. Available studies can be viewed at the bottom of the page.
Click here to learn more about Quick-Turn Studies (on-demand proficiency tests that are offered depending on lot availability). The Quick-Turn results are released within 48-72 hours whereas scheduled study reports are released within 21 days.

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes.

Proficiency Testing Schedule (2022-2023)

Drinking Water
WS22-2 March 23 - May 6, 2022
WS22-4 September 21 - November 4, 2022

WS23-2 March 22 – May 5, 2023
WS23-4 September 20 – November 3, 2023


Quick-Turn Study (on demand proficiency test that is offereddepending on lot availability)

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Chloride 20-160 mg/L
    Fluoride 1-8 mg/L
    Nitrate as NO3 1-44.3 mg/L
    Nitrate as N 3-10 mg/L
    Nitrate+nitrite as N 3-10 mg/L
    Nitrite as NO2 1-6.59 mg/L
    Nitrite as N 0.400 - 2.00
    Orthophosphate as P 0.500 - 5.50
    Phosphorus, total 0.500 - 5.50
    Sulfate 25-250 mg/L
    See All (10)

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 6

1 of 6

Supelco

Supelco

PE1401

Color - WS

Supelco

Supelco

PE1388

Demand (Low Level) - WS

Supelco

Supelco

PE1573

Total Cyanide - WS

Supelco

Supelco

PE1372

Chlorate and Chlorite - WS

Supelco

Supelco

PE1301

PNAs - WS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service