Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0820

Sigma-Aldrich

Tetramethylrhodamine-5-isothiocyanate

≥85% purity, powder

Synonym(s):

5-TRITC, Xanthylium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N3O3S
CAS Number:
Molecular Weight:
443.52
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Tetramethylrhodamine-5-isothiocyanate,

assay

≥97% (HPLC)

Quality Level

form

powder

εmax

82-98 at 540-546 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

dry ice

storage temp.

−20°C

SMILES string

CN(C)c1ccc2c(-c3ccc(cc3C([O-])=O)N=C=S)c4ccc(cc4[o+]c2c1)N(C)C

InChI

1S/C25H21N3O3S/c1-27(2)16-6-9-19-22(12-16)31-23-13-17(28(3)4)7-10-20(23)24(19)18-8-5-15(26-14-32)11-21(18)25(29)30/h5-13H,1-4H3

InChI key

IJSMFQNTEUNRPY-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service