MilliporeSigma
All Photos(1)

Documents

10032305

TR146 cell line human

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
TR-146 Cells

biological source

human mouth

description

Human squamous cell carcinoma

growth mode

Adherent

karyotype

Not specified

morphology

Polygonal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 3

This Item
SCC162SCC109
TR146 cell line human

10032305

TR146 cell line human

growth mode

Adherent

growth mode

-

growth mode

-

karyotype

Not specified

karyotype

-

karyotype

-

morphology

Polygonal

morphology

-

morphology

-

products

Not specified

products

-

products

-

receptors

Not specified

receptors

-

receptors

-

General description

The TR146 cells are appropriate for drug transport studies and they mimic normal human buccal epithelium. It serves as an oral mucosa model for barrier property studies with various molecules including tight junction proteins, aquaporins and epithelial-mesenchymal transition (EMT) markers.

Cell Line Origin

Human squamous cell carcinoma

Cell Line Description

TR146 was derived from the neck node of a 67 year-old female (primary tumour was sited in buccal mucosa). The patient had had previous radiotherapy (6,000 rads) and neck dissection. TR146 is tumourigenic in female (nu/nu) mice leading to the production of rapidly progressing tumours. Histology: well-differentiated keratinizing squamous cell carcinoma.

Application

TR146 cell line human has been used:
  • in cytotoxicity studies with an antibacterial compound, metronidazole
  • in viability and metabolic studies post blue laser light exposure,
  • as an oral epithelium-resembling cell line model for in vitro C. albicans infection studies
  • in barrier function studies using transepithelial electrical resistance (TEER)
  • for the transport studies with C-reactive protein (CRP)

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 11,14
D16S539: 13
D5S818: 10,11
D7S820: 10,12
THO1: 9
TPOX: 8,9
vWA: 15,18

Culture Medium

HAMS F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 seeding at approximately 3 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C. Saturation density of 5 x 104 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

J Jacobsen et al.
European journal of oral sciences, 107(2), 138-146 (1999-05-08)
Use of a cell culture model of a specific epithelium requires documentation of its differentiation. This study reports permeability of mannitol concurrent with a profile of differentiation markers of filter-grown TR146 cells, a cell line originating from a human buccal
Selene Mogavero et al.
Methods in molecular biology (Clifton, N.J.), 2260, 133-143 (2021-01-07)
Microbial interactions with epithelial barriers are important steps preceding disease. Infections with Candida albicans are no exception. This opportunistic fungus, commonly harmlessly residing in close proximity to human epithelia, can shift to a more pathogenic form, can invade tissues, and
Hanne Mørck Nielsen et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 16(3), 151-157 (2002-07-20)
The present study was conducted to investigate and compare the effect of pH and drug concentration on nicotine permeability across the TR146 cell culture model and porcine buccal mucosa in vitro. As a further characterization of the TR146 cell culture
J Jacobsen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 48(3), 217-224 (1999-12-28)
The dissolution rate, the toxicity and the release from chewing gum of miconazole and econazole cyclodextrin products and complexes were investigated. The dissolution rate studies showed that an amorphous miconazole hydroxypropyl-beta-cyclodextrin product gave drug supersaturation, whereas drug supersaturation was not
H T Rupniak et al.
Journal of the National Cancer Institute, 75(4), 621-635 (1985-10-01)
Four human cell lines were established from biopsy specimens of squamous cell carcinomas of the larynx (TR131 and TR138), tongue (TR126), and buccal mucosa that had infiltrated a lymph node (TR146). All 4 lines readily formed colonies on a plastic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service