MilliporeSigma
All Photos(1)

24324C

SAFC

EX-CELL® 302 Serum-Free Medium for CHO Cells

dry powder, suitable for cell culture

MDL number:
NACRES:
NA.71

description

for research or for further manufacturing use

Quality Level

form

dry powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
24326C14324C24340C
storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage condition

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable (suspension)

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500

description

for research or for further manufacturing use

description

Drug Master File available, for research or for further manufacturing use

description

for research or for further manufacturing use

description

for research or for further manufacturing use, without L-glutamine

Application

EX-CELL® 302 is a serum-free medium which has been specifically developed for the long-term growth of transformed Chinese Hamster Ovary (CHO) cells in suspension for the expression of antibodies or protein products. EX-CELL 302 is an appropriate medium for use with the DHFR- or Glutamine Synthetase, or the GS System™, because it does not contain hypoxanthine, thymidine or L-glutamine. For use with the DHFR- selection system we recommend the addition of 4 mM L-glutamine for optimal growth. Due to current regulatory concerns about the sources of raw materials, EX-CELL 302 was developed using only recombinant human proteins that have molecular weights less than 10 kDa. The total protein concentration found in EX-CELL 302 is less than 1 mg/L. Pluronic™ F68 has been added at a final concentration of 0.1% to EX-CELL 302 to protect against shear damage in sparged bioreactor systems.

Caution

Do not freeze. Protect from light. Several components of the medium are light labile and should not be exposed to light for lengthy periods of time.

Quantity

Formulated to contain 21.2 grams of powder per liter of medium.

Footnote

Ex-Cell 302 has also been used successfully with BHK-21 cells and chicken embryo-related (CER) cell lines.

Legal Information

EX-CELL is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Héla Kallel et al.
Journal of biotechnology, 95(3), 195-204 (2002-05-15)
We have carried out the adaptation of BHK-21 cells to two serum free (Ex Cell 520 and HyQ PF CHO) and three animal protein free media: Ex Cell 302, HyQ PF CHO MPS and Rencyte BHK. After a direct switch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service