MilliporeSigma
All Photos(1)

24561C

SAFC

EX-CELL® VPRO Serum-Free Medium for Retinoblast Cells

Animal-protein free, with L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture

MDL number:
NACRES:
NA.71

Quality Level

description

for research or for further manufacturing use

form

dry powder

availability

not available in Japan

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
24660C14530C14561C
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, cell culture | stem cell: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

400

Quality Level

400

description

for research or for further manufacturing use

description

for research or for further manufacturing use

description

Animal-component-free, low protein, for research or for further manufacturing use

description

for research or for further manufacturing use

availability

not available in Japan

availability

-

availability

not available in Japan

availability

-

Application

EX-CELL VPRO is an animal-protein free, serum-free medium developed for the long-term growth of Human Embryonic Retinoblast cells (PER.C6™ and related cell lines). The cells can be subcultured directly into EX-CELL VPRO from serum-free media with little or no adaptation. PER.C6 cells can be grown as suspension cultures either in shaker flasks or roller bottles, with roller bottles being the preferred culture system. Suspension cultures can be maintained, without refeeding, for approximately 10 days and can be carried for more than 20 passages with no loss of viability.

Quantity

Formulated to contain 18.5 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service