MilliporeSigma
All Photos(1)

24621C

SAFC

EX-CELL® 620-HSF Serum-Free Medium for Hybridoma Cells

Low-protein, without L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

MDL number:
NACRES:
NA.71

Quality Level

description

for research or for further manufacturing use

form

dry powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
24610C24651C24324C
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500

description

for research or for further manufacturing use

description

for research or for further manufacturing use

description

for research or for further manufacturing use

description

for research or for further manufacturing use

Application

EX-CELL 620-HSF is a low-protein (approximately 11 mg/L) serum-free medium which has been specifically developed for the long-term growth of hybridoma and related cells capable of expressing monoclonal antibodies and other protein products. The following hybridoma, lymphoma and myeloma cell lines have demonstrated sustained long-term, serum-free growth (> 15 passages) in EX-CELL 620-HSF: Sp2/0, Ag14, L243, VD-10, Hut 78, K562. Additionally, transformed Chinese Hamster Ovary (CHO) cells have demonstrated excellent growth and productivity when grown in this medium.

Quantity

Formulated to contain 17.2 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service