Skip to Content
MilliporeSigma
All Photos(1)

Documents

939B

Sigma-Aldrich

Universal Negative Control Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

form

buffered aqueous solution (red)

Quality Level

packaging

pkg of 100 mL ready to use (939B-03)
pkg of 1000 mL ready to use (939B-09)
pkg of 25 mL ready to use (939B-02)
pkg of 7 mL ready to use (939B-01)

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

The Universal Negative Control Serum reagent is used in IHC assays where both mouse and rabbit primary antibodies produce antigen specific recognition and binding. These reagents aid in the identification of cells, tissues or tissue components, which may nonspecifically bind antibodies and thereby facilitate interpretation.

Physical form

Solution containing rabbit and mouse negative serum, Tris buffer, pH 7.3-7.7, with 1% BSA and <0.1% sodium azide

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sigma-Aldrich

Sigma-Aldrich

M5905

Mouse serum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service