MilliporeSigma
All Photos(1)

Documents

AC-0361A

Sigma-Aldrich

Galectin-3 (EP412) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
345830-M434R-1246R-1
biological source

rabbit

biological source

-

biological source

rabbit

biological source

rabbit

antibody form

Ig fraction of antiserum

antibody form

-

antibody form

culture supernatant

antibody form

culture supernatant

form

buffered aqueous solution

form

lyophilized

form

buffered aqueous solution

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

technique(s)

-

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

shipped in

wet ice

shipped in

-

shipped in

wet ice

shipped in

wet ice

General description

Galectin-3 (GAL3), encoded by LGALS3, is a member of the galectin family of β-galactoside-binding lectins. GAL3 is involved in many biological process, such as cell growth and differentiation, cell-cell and cell-matrix adhesion interactions, angiogenesis, and apoptosis. It is expressed in normal breast epithelial cells, inflammatory and various malignant cells. Recent investigators have found GAL3 involved in tumor progression and metastasis, and its expression valuable in differentiating between benign and malignant thyroid neoplasms. Thyroid carcinomas are histologically classified into four subtypes: papillary, follicular, medullary, and anaplastic/undifferentiated. The papillary and follicular carcinomas are derived from follicular epithelium while the medullary subtype is derived from parafollicular cells (C cells). Anaplastic carcinoma also originates from follicular epithelium, but are highly aggressive. GAL3 was expressed in most thyroid malignancies of epithelial origin from fine-needle aspirate and open biopsies. GAL3 did not stain normal thyroid tissues or benign follicular adenomas. In a panel of 5 diagnostic markers (GAL3, FN1, CITED1, CK19, and HBME1), GAL3 demonstrated the highest sensitivity and accuracy in the differential diagnosis of benign and malignant tumors with follicular architecture. Fibronectin 1 (FN1) and GAL3 co-expression was diagnostic of carcinoma; their absence was indicative for benign lesions. GAL3 may be a useful marker for thyroid carcinomas of follicular cell origin (including papillary, follicular, and anaplastic carcinomas).

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &< 0.1%Sodium Azide

Other Notes

For a copy of the IFU and CofA contact IVDorder@ABCAM.com

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service