MilliporeSigma
All Photos(1)

Documents

C1370

Sigma-Aldrich

COT (30-397), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Synonym(s):
EST, FLJ10486, MAP3K8, TPL2, c-COT
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

860-1164 nmol/min·mg

mol wt

~70 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP3K8(1326)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
M0324M1198M8822
vibrant-m

M0324

MEKK2, active, GST tagged human

vibrant-m

M1198

MEK2, active, GST tagged human

vibrant-m

M8822

MEK1, active, GST tagged human

specific activity

860-1164 nmol/min·mg

specific activity

192-260 nmol/min·mg

specific activity

190-258 nmol/min·mg

specific activity

276-374 nmol/min·mg

Gene Information

human ... MAP3K8(1326)

Gene Information

human ... MAP3K2(10746)

Gene Information

human ... MAP2K2(5605)

Gene Information

human ... Map2k1(26395)

mol wt

~70 kDa

mol wt

~115 kDa

mol wt

~71 kDa

mol wt

~69 kDa

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

Biochem/physiol Actions

COT is an oncogene that can activate both the MAP kinase and JNK kinase pathways. COT activates I?B kinases and induces the nuclear production of NF-?B. C-terminal catalytic domain of KSR2 associates with COT and KSR2 can negatively regulates the kinase activity of COT in vitro. Co-transfection of KSR2 with COT in cells lead to reduced COT-mediated ERK activation and COT-induced IL8 production in a dose-dependent manner. COT is one of the MAP kinase kinase kinases that regulates the ERK1/ERK2 pathway in response to IL-1. Blockage of expression of COT results in failure of IL-1 to induce an increase in IL-8 and MIP-1betamRNA levels.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Cristina Rodríguez et al.
Cellular signalling, 18(9), 1376-1385 (2005-12-24)
Cot is one of the MAP kinase kinase kinases that regulates the ERK1/ERK2 pathway under physiological conditions. Cot is activated by LPS, by inducing its dissociation from the inactive p105 NFkappaB-Cot complex in macrophages. Here, we show that IL-1 promotes
Padma L Channavajhala et al.
The Journal of biological chemistry, 278(47), 47089-47097 (2003-09-17)
Kinase suppressor of Ras (KSR) is an integral and conserved component of the Ras signaling pathway. Although KSR is a positive regulator of the Ras/mitogen-activated protein (MAP) kinase pathway, the role of KSR in Cot-mediated MAPK activation has not been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service