MilliporeSigma
All Photos(1)

Documents

MEF Ulk1 -/- Ulk2 -/- (DKO) (SV40)

14050802, from mouse embryo

Sign Into View Organizational & Contract Pricing

biological source

mouse embryo

description

Mouse embryonic fibroblast, autophagy, Ulk1 -/- Ulk2 -/-, double knockout, SV40-immortalised

growth mode

Adherent

morphology

Fibroblast

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Description

Primary mouse embryonic fibroblasts (MEFs) were isolated from the embryos of a pregnant female Ulk1-/- Ulk2-/+ mouse at day 13.5 post coitus. The embryos were genotyped to identify those that were Ulk1 -/- Ulk2 -/- and the MEFs that were isolated and cultured were immortalised with SV40 large T-antigen.

Culture Medium

DMEM + 10% FCS +2mM Glutamine

Subculture Routine

Trypsinize with 0.05% Trypsin/EDTA and inoculate new cultures at 2 x 104 per cm2. Cultures must be incubated in a humidified 5% CO2/95% air incubator at 37oC.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service