FL (HeLa derivative)
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human cervix
packaging
tube of 5 μg 90111910-DNA-5UG
pkg of vial of cells 90111910-1VL
growth mode
Adherent
karyotype
Modal no. 71
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human cervix carcinoma
Cell Line Description
Originally derived from normal amniotic tissue. Have been used and studied extensively including virus susceptibility, tumorigenicity and morphology. Contains HeLa marker chromosomes and exhibits type A G6PD. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. The cells are capable of growth in a variety of media supplemented with sera from several species. Ethnicity: Black.
Application
Virus, tumourigenicity and chromosome studies
Culture Medium
EMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service