MilliporeSigma
All Photos(1)

Documents

CLLS1211

Sigma-Aldrich

MCF10A CELLS PVRL4 (-/-)

Sign Into View Organizational & Contract Pricing

NACRES:
NA.81

biological source

human female mammary glands (Source Disease: Fibrocystic Disease)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CLLS1069CLLS1046CLLS1042
Sigma-Aldrich

CLLS1211

MCF10A CELLS PVRL4 (-/-)

Sigma-Aldrich

CLLS1069

MCF10A CELLS APC (-/-)

Sigma-Aldrich

CLLS1046

MCF10A CELLS PTEN (-/-)

Sigma-Aldrich

CLLS1042

MCF10A CELLS CDH1 (-/-)

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

Gene Information

human ... PVRL4(81607), nectin-4(81607)

Gene Information

human ... APC(324)

Gene Information

human ... PTEN(5728)

Gene Information

human ... CDH1(1105)

OMIM accession no.

609607

OMIM accession no.

611731

OMIM accession no.

601728

OMIM accession no.

602118

General description

MCF10A CELLS PVRL4 (-/-) are pre-neoplastic mammary epithelial cells from a human adult female with a ZFN modification.

This cell line was derived from ATCC Catalog No. CRL-10317.

Application

Media Renewal changes two to three times per week.

Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.

Subculture Ratio: approx. 1:3-1:8

The base medium for this cell line is Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F-12, Cat. No. 51448C. To make the complete growth medium, add the following components to the base medium: horse serum, Cat. No. H1270, to a final concentration (v/v) of 5%; cholera toxin, Cat. No. C8052, to a final concentration of 1 ng/mL; human insulin, Cat. No. I9278, to a final concentration of 10 ug/mL; epidermal growth factor, Cat. No. E9644, to a final concentration of 10 ng/mL; and hydrocortisone, Cat. No. H6909, to a final concentration of 0.5 ug/mL.

Cell freezing medium - DMSO 1X, Cat. No. C6164.

Features and Benefits

These MCF10A cells are adherent with a doubling time of approx. 20 hours

Zinc finger nuclease (ZFN) knockout on chromosome 1q22-q23.2

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • MCF10A CONTROL CELLS 1 vial

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service