MilliporeSigma
All Photos(7)

Documents

CWA-1034

Sigma-Aldrich

Anti-Human CD63 (MEM-259) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MEM-259, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 25.64 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

technique(s)

flow cytometry: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD63(967)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CWA-1021CWA-1062CWA-1026
Gene Information

human ... CD63(967)

Gene Information

mouse ... IL2RA(3559)

Gene Information

human ... ITGB1(3688)

Gene Information

mouse ... FCGR1A(2209)

clone

MEM-259, monoclonal

clone

BC96, monoclonal

clone

MEM-101A, monoclonal

clone

10.1, monoclonal

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

product line

ColorWheel®

product line

ColorWheel®

product line

ColorWheel®

product line

ColorWheel®

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone MEM-259 is a mouse monoclonal antibody that detects human CD63.

Immunogen

HPB-Acute lymphocytic leukemia cells.

Application

Quality Control Testing:

Evaluated by Flow Cytometry in K562 cells.

Flow Cytometry Analysis: Staining of one million human K562 cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1034, Anti-Human CD63 (MEM-259) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® activated Phycoerythrin (PE) Dye or an equivalent amount of PE-conjugated Mouse IgG1 Isotype Control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

CD63 antigen (UniProt: P08962; also known as Granulophysin, Lysosomal-associated membrane protein 3, LAMP-3, Melanoma-associated antigen ME491, OMA81H, Ocular melanoma-associated antigen, Tetraspanin-30, Tspan-30, CD63) is encoded by the CD63 (also known as MLA1, TSPAN30) gene (Gene ID: 967) in human. CD63 is a multi-pass membrane protein of the tetraspan family that is found on endosome, lysosome, and plasma membranes. CD63 has been detected in platelets, dysplastic nevi benign moles, radial growth phase primary melanomas, hematopoietic cells, and in tissue macrophages. In melanoma cells, it is involved in their motility and adhesion. CD63 also plays a role in the adhesion of leukocytes onto endothelial cells. It is reported to play a role in the activation of ITGB1 and integrin signaling, leading to the activation of AKT, FAK/PTK2 and MAP kinases and promote cell survival, reorganization of the actin cytoskeleton, cell adhesion, spreading and migration. CD63 is a highly N-glycosylated protein with three asparagine glycosylation sites (aa 130, 150, 172) and its ribophorin II (RPN2)-mediated glycosylation has been linked to breast cancer. Overexpression of CD63 has been observed in esophageal cancer that is negatively correlated with tumor stage and lymph node metastasis. Lack of expression of CD63 in platelets has been observed in a patient with Hermansky-Pudlak syndrome (HPS), an autosomal recessive disorder that is characterized by oculocutaneous albinism, bleeding due to platelet storage pool deficiency, and lysosomal storage defects. (Ref.: Lai, X., et al. (2017). Oncol. Let. 13(6); 4245-4251; Tominaga, N., et al. (2014). Mol. Cancer 13; 134; Smith, M., et al. (1997). Melanoma Res. 7 (Suppl. 2), 163-170).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Related product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Articles

Harness the flexibility of ColorWheel® flow cytometry antibodies and dyes to unlock freedom in your flow cytometry multiplex assay design or multicolor panel design and simplify your multiplex flow cytometry workflows.

Protocols

Experience simplicity in your flow cytometry workflow with the 3-Step ColorWheel® Flow Cytometry Reagent Preparation protocol. With less than 5 minutes of hands-on time, see how simple it is to create your own optimal flow cytometry reagents with ColorWheel® flow cytometry antibodies and dyes.

View ColorWheel® protocol steps for flow cytometry analysis when using ColorWheel® antibodies with ColorWheel® dyes including antibody preparation, PBMC sample preparation, cell surface staining, and intracellular (cytoplasmic) staining.

Related Content

Unlock freedom in your flow cytometry assay design with ColorWheel® flow cytometry antibodies and dyes. The ColorWheel® flow cytometry portfolio was designed with simplicity and flexibility in mind to help streamline your flow cytometry workflow without compromising on quality.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service