Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU046421

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Traf6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTCCCTGACGGTAAAGTGCCCAAATAAAGGCTGTTTGCAAAAGATGGAACTGAGACATCTCGAGGATCATCAAGTACATTGTGAATTTGCTCTAGTGAATTGTCCCCAGTGCCAACGTCCTTTCCAGAAGTGCCAGGTTAATACACACATTATTGAGGATTGTCCCAGGAGGCAGGTTTCTTGTGTAAACTGTGCTGTGTCCATGGCATATGAAGAGAAAGAGATCCATGATCAAAGCTGTCCTCTGGCAAATATCATCTGTGAATACTGTGGTACAATCCTCATCAGAGAACAGATGCCTAATCATTATGATCTGGACTGCCCAACAGCTCCAATCCCTTGCACATTCAGTGTTTTTGGCTGTCATGAAAAGATGCAGAGGAATCACTTGGCACGACACTTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melinda E Varney et al.
The Journal of experimental medicine, 212(11), 1967-1985 (2015-10-16)
TRAF-interacting protein with forkhead-associated domain B (TIFAB) is a haploinsufficient gene in del(5q) myelodysplastic syndrome (MDS). Deletion of Tifab results in progressive bone marrow (BM) and blood defects, including skewed hematopoietic stem/progenitor cell (HSPC) proportions and altered myeloid differentiation. A
Frank Secreto et al.
Leukemia & lymphoma, 55(8), 1884-1892 (2013-11-12)
B-cell activating factor-receptor (BAFF-R) is the primary BAFF receptor that is responsible for promoting B-cell development and survival. Malignant B-cells exploit the BAFF/BAFF-R system, and high serum BAFF levels or genetic alterations in BAFF receptors have been found in B-cell
Domenico Somma et al.
Journal of immunology (Baltimore, Md. : 1950), 194(7), 3286-3294 (2015-02-25)
IL-17 is a proinflammatory cytokine that promotes the expression of different cytokines and chemokines via the induction of gene transcription and the posttranscriptional stabilization of mRNAs. In this study, we show that IL-17 increases the half-life of the Zc3h12a mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service