MilliporeSigma
All Photos(1)

Documents

GB11

Sigma-Aldrich

“Good” buffers

Sign Into View Organizational & Contract Pricing

NACRES:
NA.25

 

(1) 6, (2) 8

cation traces

heavy metals: <5 ppm

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PPB0231.094331.09436
Sigma-Aldrich

Sigma-Aldrich

GB11

“Good” buffers

Tris buffer Pre-set, pH 8.1, pHast Pack&#8482;, powder

Sigma-Aldrich

PPB023

Tris buffer

Buffer solution (citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 2.00 (20&#176;C) Certipur&#174;

Supelco

1.09433

Buffer solution

Buffer solution (citric acid/sodium hydroxide) traceable to SRM from NIST and PTB pH 5.00 (20&#176;C) Certipur&#174;

Supelco

1.09436

Buffer solution

 

(1) 6, (2) 8

 

-

 

-

 

-

General description

Good′s buffers are a set of 20 buffers that were prepared by Good et al. They are useful in biological and biochemical research. These buffers, which are zwitter-ionic compounds, have a pKa value between 6 and 8. Good′s buffers have low toxicity, are highly water-soluble, stable against pH changes, have hydrolytic and enzymatic stability and have less effects on biochemical reactions.

Packaging

11 Buffers in quantities indicated

Kit Components Also Available Separately

Product No.
Description
SDS

  • B9879BES 25 gSDS

  • B3876BICINE 100 gSDS

  • C2632CAPS 100 gSDS

  • E9502EPPS 100 gSDS

  • H3375HEPES 25 gSDS

  • M8250MES 25 gSDS

  • M1254MOPS 100 gSDS

  • P6757PIPES 25 gSDS

  • T5130TAPS 100 gSDS

  • T1375TES 100 gSDS

  • T0377TRICINE 100 gSDS

See All (11)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 6

1 of 6

HEPBS &#8805;99% (titration)

Sigma-Aldrich

H6903

HEPBS

TAPS sodium salt &#8805;99% (titration)

Sigma-Aldrich

T0647

TAPS sodium salt

BES sodium salt &#8805;99% (titration)

Sigma-Aldrich

B2891

BES sodium salt

Tricine &#8805;99% (titration)

Sigma-Aldrich

T0377

Tricine

Joachim W.
Handbook of Ion Chromatography (2016)
Álvaro de Mingo Pulido et al.
Cancer cell, 33(1), 60-74 (2018-01-10)
Intratumoral CD103
Hydrogen ion buffers for biological research.
N E Good et al.
Biochemistry, 5(2), 467-477 (1966-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service