MilliporeSigma
All Photos(1)

Documents

I0282

Sigma-Aldrich

Anti-α-Internexin antibody, Mouse monoclonal

clone 2E3, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing

Synonym(s):
Anti-66 kDa Neurofilament Subunit, Anti-INA, Anti-NF-66
MDL number:
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

2E3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~66 kDa by SDS-PAGE

species reactivity

feline, rat, bovine, human, pig

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5 μg/mL using rat brain cytosolic extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INA(9118)
rat ... Ina(24503)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
N2787SAB4200740L7391
conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody form

purified immunoglobulin

antibody form

purified from hybridoma cell culture

antibody form

purified from hybridoma cell culture

species reactivity

feline, rat, bovine, human, pig

species reactivity

human, mouse, rat, zebrafish

species reactivity

-

species reactivity

zebrafish, chicken, rat, human, bovine, mouse

clone

2E3, monoclonal

clone

RMO44, monoclonal

clone

NN18, monoclonal

clone

LIS1-338, monoclonal

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

Monoclonal Anti-α -Internexin (mouse IgG1 isotype) is derived from the 2E3 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from mice immunized with the full-length recombinant rat α -internexin.
The gene INA (α-Internexin) encodes a neuronal intermediate filament (IF) protein that is a type IV IF. The gene is mapped to human chromosome 10. The encoded protein has a molar mass of 55.4kDa and spans a length of 499 amino acids. It is expressed in the neurons along with the NF (neurofilament) triplet proteins, NF-L (light), NF-M (medium), NF-H (heavy).

Immunogen

full-length recombinant rat α-internexin.

Application

Anti-α-Internexin antibody, Mouse monoclonal has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunohistochemistry
  • immunocytochemistry

Biochem/physiol Actions

The expression of α-Internexin precedes the expression of NF triplet proteins during the differentiation of neurons. Its expression is relatively low when compared to the the NF proteins in the adult brain. It co-assembles with the NF triplet proteins. Overexpression of α-internexin in transgenic mouse model has been found to cause abnormal neurofilamentous accumulations and motor coordination deficits. It has been found to be present in cytoplasmic inclusions linking it to NIFID (neuronal intermediate filament inclusion disease) and other neurological diseases with pathological accumulations of IFs (intermediate filaments). α-Internexin may be useful as a biomarker for pancreatic neuroendocrine tumor aggressiveness and prognosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Nestin in immature embryonic neurons affects axon growth cone morphology and Semaphorin3a sensitivity
Bott CJ, et al.
Molecular Biology of the Cell, 30(10), 1214-1229 (2019)
Bei Liu et al.
The Journal of clinical endocrinology and metabolism, 99(5), E786-E795 (2014-02-04)
We aimed to test whether α-internexin could be a molecular biomarker of tumor aggressiveness and prognosis in pancreatic neuroendocrine tumors (PNETs). Using immunohistochemical staining and Western blot, we detected the expression of α-internexin in 350 tumors from 343 patients, of
C J Bott et al.
Molecular biology of the cell, 30(10), 1214-1229 (2019-03-07)
Correct wiring in the neocortex requires that responses to an individual guidance cue vary among neurons in the same location, and within the same neuron over time. Nestin is an atypical intermediate filament expressed strongly in neural progenitors and is
Nigel J Cairns et al.
The American journal of pathology, 164(6), 2153-2161 (2004-05-27)
Neuronal intermediate filament (IF) inclusion disease (NIFID) is a novel neurological disease of early onset with a variable clinical phenotype including frontotemporal dementia, pyramidal, and extrapyramidal signs. Pathologically, in affected areas, there is neuronal loss, astrocytosis, and neuronal intracytoplasmic aggregates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service