MAK264
High Sensitivity Triglyceride Fluorometric Assay Kit
sufficient for 100 fluorometric tests
Synonym(s):
Triglyceride Quantification Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
application(s)
cosmetics
food and beverages
detection method
fluorometric
relevant disease(s)
neurological disorders; cardiovascular diseases
storage temp.
−20°C
General description
Triglycerides (TG) are the main constituents of vegetable oil, animal fat, LDL and VLDL, and play an important role as transporters of fatty acids as well as an energy source. TG is broken down into fatty acids and glycerol, after which both can serve as substrates for energy producing and metabolic pathways. High blood levels of TG are implicated in atherosclerosis, heart disease and stroke as well as in pancreatitis.
Features and Benefits
Compatible with high-throughput handling systems.
Suitability
Suitable for measuring triglyceride levels in serum, plasma, saliva, and other biological fluids, and tissue and cell culture samples.
Principle
The High Sensitivity Triglyceride Assay Kit is suitable for measuring triglyceride levels in samples that contain reducing substances that may interfere with oxidase-based assays. In this assay, TG is hydrolyzed with lipase to glycerol and fatty acids. The glycerol is used in a coupled enzyme reaction, which results in a fluorometric product (λex = 535 nm/λem = 587 nm), proportional to the amount of triglycerides present. This high-throughput, adaptable assay kit is simple and sensitive with the capability to detect less than 0.4 μM of triglycerides in a variety of samples.
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service