MilliporeSigma
All Photos(1)

Documents

MAK270

Sigma-Aldrich

High Sensitivity Free Glycerol Fluorometric Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing

NACRES:
NA.25

application(s)

cosmetics
environmental
food and beverages

detection method

fluorometric

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
FG0100MAK117MAK261
Free Glycerol Determination Kit 1 kit sufficient for 1000 reactions

FG0100

Free Glycerol Determination Kit

Glycerol Assay Kit sufficient for 200 colorimetric or fluorometric tests

MAK117

Glycerol Assay Kit

Glycine Assay Kit (Fluorometric) sufficient for 100 fluorometric tests

MAK261

Glycine Assay Kit (Fluorometric)

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

application(s)

cosmetics
environmental
food and beverages

application(s)

-

application(s)

cosmetics
environmental
food and beverages

application(s)

-

General description

Glycerol, also referred to as glycerin or glycerine, is a 3 carbon sugar alcohol that forms the backbone of fatty acids. It is a central component for synthesis of all lipids, and acts as a backbone for triglycerides and phospholipids, which play an important role in cell membrane structure. Due to its low toxicity, glycerol is widely used in pharmaceutical, food and cosmetic formulations and is the main waste by-product of biodiesel production via transesterification

Suitability

Suitable for use with serum, plasma, and other biological fluids, and tissue and cell culture samples.

Principle

The High Sensitivity Free Glycerol Assay Kit is suitable for measuring trace amounts of glycerol in samples, which contain reducing substances that may interfere with oxidase-based assays. This assay employs coupled enzyme reaction, which results in a fluorometric product (lex = 535 nm/lem = 587 nm), proportional to the amount of glycerol present. This assay kit is simple and sensitive with the capability to detect <40 pmole glycerol in a variety of samples.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service