Skip to Content
MilliporeSigma

MAK639

Peroxide Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Peroxide Detection Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116104
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 100 colorimetric tests

detection method

colorimetric

storage temp.

−20°C

General description

Peroxide(hydrogen peroxide H2O2) is one of the key reactiveoxygen species formed under oxidative stress conditions. High levels ofperoxide formation have been linked to pathological conditions such as aging,asthma, diabetes, atherosclerosis, cataracts, inflammatory arthritis andneurodegenerative diseases.

Application

Suitable for the direct assay of H2O2 and organic peroxides in biological samples (e.g. serum, citrate-plasma, urine, cell lysate, culture medium) and the determination of the effects of drugs on peroxide metabolism.

Biochem/physiol Actions

The improvedperoxide assay kit is designed to measure peroxide concentration in biologicalsamples without any pretreatment. This method utilizes the chromogenic Fe3+‑xylenolorange reaction, in which a purple complex is formed when Fe2+provided in the reagent is oxidized to Fe3+ by peroxides present inthe sample. The intensity of the color, measured at 540‑610 nm, is an accuratemeasure of the peroxide level in the sample. The optimized formulationsubstantially reduces interference by substances in the raw samples.

This assayhas a detection limit of 0.54 µM (18.4 ng/mL) and linear up to 30 µM (1,020ng/mL) H2O2 in 96-well plate assay.


Still not finding the right product?

Explore all of our products under Peroxide Assay Kit


Storage Class

10 - Combustible liquids

wgk

nwg



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content