MilliporeSigma
All Photos(1)

Documents

SAB5700290

Sigma-Aldrich

Anti-Phospho-EGFR-Y998 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Synonym(s):
EGFR, ERBB, ERBB1, HER1, NISBD2, PIG61, mENA

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

134
44
69
77

species reactivity

human

concentration

1.25 mg/ml

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr998)

Gene Information

human ... EGFR(1956)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB5700292SAB5700289SAB5700347
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

mol wt

134, 69, 44, 77

mol wt

134, 44, 69, 77

mol wt

134, 44, 69, 77

mol wt

134, 44, 69, 77

technique(s)

western blot: 1:500-1:2000

technique(s)

western blot: 1:500-1:2000

technique(s)

western blot: 1:500-1:2000

technique(s)

immunohistochemistry: 1:50-1:100, immunoprecipitation (IP): 1:50-1:100, western blot: 1:1000-1:2000

UniProt accession no.

P00533

UniProt accession no.

P00533

UniProt accession no.

P00533

UniProt accession no.

P00533

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer.

Immunogen

A synthetic phosphorylated peptide around Y998 of human EGFR (NP_005219.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service