Z744129
eXTReme™ 2 Seal
polypropylene, W × L 3.083 in. × 5.53 in.
Synonym(s):
eXTReme™ 2 Seal, PCR film, PCR tape, adhesive sealing film, adhesive sealing tape, microplate sealing film, microplate sealing tape, sealing film, sealing tape
About This Item
material
polypropylene
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Excel Scientific XTR2-100
parameter
-20-+120 °C temp. range
technique(s)
PCR: suitable
W × L
3.083 in. × 5.53 in.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Certified DNase- & RNase- free
- Next-generation adhesive formula provides better-than-ever sample protection with improved evaporation rates
- Standard cut features chamfered corners to eliminate plate overhang for robotics applications and tight storage conditions
- Standard cut is ideal for use with raised-rim and nonskirted PCR plates
- Large cut is designed for greater plate coverage and easier positioning of the film to the plate
- Large cut is recommended for use with flat-top, semiskirted and full-skirted PCR plates as well as deepwell plates NOT recommended for qPCR
Application
Features and Benefits
- Removes cleanly from plate surface
- Minimizes evaporation
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service