Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O8, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~28.32 and 20.66 kDa
observed mol wt ~30 and 18 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Methyltransferase-like protein 7A (UniProt: Q9H8H3 and computationally mapped F8VQX6, Q9H8H3, also known as AAM-B, METTL7A) is encoded by the METTL7A gene (Gene ID: 25840) in human. METTL7A is a ubiquitously expressed lipid droplets-associated protein that displays high expression in liver and thyroid tissues. It is synthesized with a signal peptide (aa 1-29) and is an integral membrane protein anchored into the endoplasmic reticulum (ER) membrane. It contains N and C-terminal domains, juxtamembrane region, and a putative central catalytic domain for methyltransferase activity. The N-terminal regions contains a hydrophobic sequence of 28 amino acids that is shown to be essential for its insertion into ER. It also helps recruit other cellular protein to lipid droplets for the formation of functional organelles. METTL7A is reported to be essential for the assembly and release stages of hepatitis C (HCV) life cycle but is not essential for the replication step of virus. Knockdown of METTL7A is shown to impair HCV propagation. METTL7A gene is reported to be downregulated in thyroid cancer. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhou, S., et al., (2017). Oncotarget. 8(21); 34652-34660; Park, EM., et al. (2015). PLoS One. 10(7); e0132839).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects human Methyltransferase-like protein 7A (METTL7A). It targets an epitope within 21 amino acids from the C-terminal region and detects both isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human Methyltransferase-like protein 7A (METTL7A).

Application

Quality Control Testing

Evaluated by Western Blotting in HT-1376 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Methyltransferase-like protein 7A (METTL7A) in HT-1376 cell lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound METTL7A peptide with a KD of 1.1 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected METTL7A in human testis tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected METTL7A in HepG2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-METTL7A, clone 1O8 ZooMAb®, Cat. No. ZRB1185, is a rabbit recombinant monoclonal antibody that targets METTL7A and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service