ZRB1528
Anti-GLTSCR1L Antibody, clone 1L13 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
BRD4-interacting chromatin-remodeling complex-associated protein-like;Glioma tumor suppressor candidate region gene 1 protein-like
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1L13, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 115.08 kDa
observed mol wt ~110 kDa
purified by
using Protein A
species reactivity
human, mouse
species reactivity (predicted by homology)
nonhuman primates
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal half
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... BICRAL(23506)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in HCT116 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GLTSCR1L in HCT116 cell lysate.
Tested applications
Western Blotting Analysis (WB): A 1:1,000 dilution from a representative lot detected GLTSCR1L in NIH3T3, A431, Raw264.7, and Jurkat cell lysates.
Western Blotting Analysis: A representative lot of this antibody detected GLTSCR1L in mononuclear primary cell cultures from bone marrow, spleen plus human PlatE cells, and LNCaP cells.
Flow Cytometry Analysis (FC): 1 μg from a representative lot detected GLTSCR1L in one million Jurkat cells.
Affinity Binding Assay: A representative lot of this antibody bound GLTSCR1L peptide with a KD of 3.2 x 10-6 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Evaluated by Western Blotting in HCT116 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GLTSCR1L in HCT116 cell lysate.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service