MilliporeSigma
All Photos(16)

Documents

ZRB1713

Sigma-Aldrich

Anti-p-ATF-2-Thr71 Antibody, clone 1I8 ZooMAb® Rabbit Monoclonal

enhanced validationgreener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Synonym(s):
Cyclic AMP-dependent transcription factor ATF-2;cAMP-dependent transcription factor ATF-2;Activating transcription factor 2;Cyclic AMP-responsive element-binding protein 2;CREB-2;cAMP-responsive element-binding protein 2;HB16;cAMP response element-binding

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I8, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 54.54 kDa
observed mol wt ~45 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pThr71)

Gene Information

human ... ATF2(1386)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ZRB2051ZRB1721ZRB1655
species reactivity

human

species reactivity

rat, human, mouse

species reactivity

human

species reactivity

mouse, human, rat

Gene Information

human ... ATF2(1386)

Gene Information

human ... ATF3(467)

Gene Information

human ... EGR1(1958)

Gene Information

human ... ELF5(2001)

clone

1I8, recombinant monoclonal

clone

1G12, recombinant monoclonal

clone

1F22, recombinant monoclonal

clone

1K15, recombinant monoclonal

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I8 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects ATF-2 phosphorylated on Threonine 71.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphothreonine 71 in human ATF-2.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from serum starved and EGF treated HepG2 cells.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected ATF-2 phosphorylated on Thr 71 in lysate from overnight serum starved HepG2 cells treated with recombinant human EGF (200 ng/mL; 30 min), but not in lysates from untreated cells.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ATF-2 phosphorylated on Threonine 71 in lysate from HeLa cells treated with Anisomycin (25 g/mL; 30 min).

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ATF-2 phosphorylated on Threonine 71 in HepG2 cells treated with EGF (200 ng/mL; 30 min).

Affinity Binding Assay: A representative lot of this antibody bound Phospho-ATF-2-Thr71 peptide with a KD of 1.6 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ATF-2 phosphorylated on Threonine 71 in human prostate cancer tissue sections.

Peptide Inhibition Assay: Target band detection in HepG2 cell lysate was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in lysate from serum starved and EGF treated HepG2 cells.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected ATF-2 phosphorylated on Thr 71 in lysate from overnight serum starved HepG2 cells treated with recombinant human EGF (200 ng/mL; 30 min), but not in lysates from untreated cells.

Target description

Cyclic AMP-dependent transcription factor ATF-2 (UniProt: P15336; also known as cAMP-dependent transcription factor ATF-2, Activating transcription factor 2, Cyclic AMP-responsive element-binding protein 2, CREB-2, cAMP-responsive element-binding protein 2, HB16, cAMP response element-binding protein CRE-BP1) is encoded by the ATF2 (also known as CREB2, CREBP1) gene (Gene ID: 1386) in human. ATF2 is a member of the activating protein-1 (AP1) transcription factor family that regulates the transcription of various genes, including those involved in anti-apoptosis, cell growth, and DNA damage response. Dependent on its binding partner, it binds to cAMP response element (CRE) consensus sequences (5′-TGACGTCA-3′) or to AP-1 consensus sequences (5′-TGACTCA-3′). ATF2 is reported to shuttle between the cytoplasm and the nucleus and its heterodimerization with Jun is essential for its nuclear localization. It is localized in the cytoplasm under conditions of cellular stress and in disease states. It can also localize at the mitochondrial outer membrane in response to genotoxic stress. In the cytoplasm, it is shown to interact with and perturb hexokinase 1 and VDAC1-containing complexes at the mitochondrial outer membrane and impair mitochondrial membrane potential that leads to mitochondrial leakage and promotes cell death. Under basal conditions, ATF2 is auto-inhibited through intramolecular contact between the zinc finger (aa 25-49) and transactivation domain (TAD)-containing N-terminal domain and C-terminal basic leucine zipper (bZIP; aa 352-415) domain. Following activation by kinases that phosphorylate the N-terminal region, it is activated and translocates to the nucleus to modulate gene expression. Its phosphorylation at threonine 69 and 71 by MAP kinases, in response to external stimuli, increases its transcriptional activity. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bhoumik, A., et al. (2005). Mol. Cell. 18(5); 577-587; Ouwens, DM., et al. (2002). EMBO J. 21(14); 3782-3793).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service