Skip to Content
MilliporeSigma

ZRB3207

Anti-α1-Antitrypsin Antibody, clone 1K23 ZooMAb® Rabbit Monoclonal

greener alternative

Greener Alternative

enhanced validation

Enhanced Validation

recombinant, expressed in HEK 293 cells

Synonym(s):

Alpha-1-antiproteinase, Alpha-1-antitrypsin, Serpin A1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Conjugate:
unconjugated
Clone:
1K23, recombinant monoclonal
Application:
IF, IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1K23, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 46.74 kDa, observed mol wt ~49/57 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SERPINA1(5265)

General description

Alpha-1-antitrypsin (UniProt: P01009; also known as Alpha-1 protease inhibitor, Alpha-1-antiproteinase, Serpin A1) is encoded by the SERPINA1 (also known as AAT, PI) gene (Gene ID: 5265) in human. The protein encoded by this gene belongs to the serpin family and is a strong serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Its pivotal role is to protect lower respiratory tract against proteolytic destruction by human leukocyte elastase. SERPINA protein is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to produce the mature protein. It is synthesized mainly in the liver and secreted to the bloodstream. It is predominantly found in the plasma, extracellular space, and extracellular matrix. Three Isoforms of -1-antitrypsin have been described that are produced by alternative splicing. Mutation in AAT gene will reduce the serum concentration of -1-antitrypsin and hence increase the risk of emphysema or liver disease. Recent studies suggest that AAT also exhibits anti-inflammatory and immunomodulatory functions, expanding its relevance beyond traditional protease inhibition. The complex formation of fatty acids or lipoproteins with native or modified AAT impacts its protease inhibitory action as well as its anti-inflammatory properties. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Segal, L. et al., (2022). Cell Immunol. 375:104528; O′Brien, M.E., et al. (2022). Int. J. Mol. Sci. 23(5);2441).
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the internal region of human 1-Antitrypsin.

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected 1-Antitrypsin in HepG2 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected 1-Antitrypsin in human plasma and in human liver tissue lysate.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected 1-Antitrypsin in human liver tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected 1-Antitrypsin in human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1K23 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects 1-Antitrypsin. It targets an epitope within 23 amino acids from the Internal region.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

0.3 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library