MilliporeSigma
All Photos(1)

Documents

47058

Supelco

WSFA-4

0.1 wt. % each component in deionized water, qualitative standard

Sign Into View Organizational & Contract Pricing

grade

qualitative standard

CofA

current certificate can be downloaded

packaging

pkg of 1 × 5 mL

concentration

0.1 wt. % each component in deionized water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

storage temp.

2-30°C

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
4705646985-UCRM47792
Supelco

Supelco

47058

WSFA-4

Supelco

Supelco

47056

WSFA-2

format

multi-component solution

format

multi-component solution

format

multi-component solution

format

multi-component solution

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

packaging

pkg of 1 × 5 mL

packaging

pkg of 1 × 5 mL

packaging

pkg of 100 mL

packaging

ampule of 1 mL

concentration

0.1 wt. % each component in deionized water

concentration

0.1 wt. % each component in deionized water

concentration

0.01 meq/mL each component in deionized water

concentration

10 mg/mL in dichloromethane

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

General description

WSFA-4 may be used for the qualitative determination of free fatty acids in aqueous solutions. It is recommended that it should be purchased from chemical manufacturers with knowledge in the preparation, handling, storage, and shipment of volatile analytes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acetic acid
    Butyric acid
    Isobutyric acid
    Isovaleric acid
    2-Methylbutyric acid
    Propionic acid
    Valeric acid

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service