MilliporeSigma
All Photos(1)

Documents

47914

Supelco

EPA 625/CLP Pesticide Mix

2000 μg/mL each component in hexane: toluene (1:1), PESTANAL®, certified reference material

Sign Into View Organizational & Contract Pricing

grade

certified reference material

Quality Level

agency

US EPA/CLP 625 (Mar ′90, Dec ′90, Aug ′91 Statement of Work)

product line

PESTANAL®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in hexane: toluene (1:1)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

EPA 625/CLP Pesticide Mix has been used as a reference standard for instrument calibration, recovery determination and quantification of organochlorine pesticides (OCPs) and polychlorinated biphenyls (PCBs) in marine environment by gas chromatography - mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Aldrin
    α-BHC
    β-BHC
    Lindane
    δ-BHC
    4,4′-DDT
    Dieldrin
    α-Endosulfan
    β-Endosulfan
    Endosulfan sulfate
    Endrin
    Endrin aldehyde
    Heptachlor
    Heptachlor exo-epoxide
    See All (14)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service