MilliporeSigma
All Photos(1)

Documents

91326

Supelco

1-Octanesulfonic acid sodium salt

analytical standard

Linear Formula:
C8H17O3SNa
CAS Number:
Molecular Weight:
216.27
MDL number:

grade

analytical standard

Assay

97.5-102.5% (T)

form

powder

shelf life

limited shelf life, expiry date on the label

impurities

≤2.0% water

SMILES string

[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
O0133RDD0321.18307
grade

analytical standard

grade

-

grade

-

grade

-

form

powder

form

-

form

-

form

solid

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

impurities

≤2.0% water

impurities

≤0.005% Phosphorus (P), ≤0.1% Insoluble matter

impurities

-

impurities

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

Application

Ion-pairing reagent for HPLC analysis of peptides and proteins.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (Example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service