Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O8381-1AMP

Supelco

NHI-D FAME mix

analytical standard, ampule of 100 mg (Neat)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

form

solid

CofA

current certificate can be downloaded

packaging

ampule of 100 mg (Neat)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

ester (methy
methyl)

shipped in

dry ice

storage temp.

−20°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Methyl myristate 11.8 wt. %
    Methyl oleate 44.6 wt. %
    Methyl palmitate 23.6 wt. %
    Methyl palmitoleate 6.9 wt. %
    Methyl stearate 13.1 wt. %

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service