Skip to Content
MilliporeSigma

Y0002429

Cannabidiol CRS

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cannabidiol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
MDL number:
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

BP

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

InChI

1S/C21H30O2/c1-5-6-7-8-16-12-19(22)21(20(23)13-16)18-11-15(4)9-10-17(18)14(2)3/h11-13,17-18,22-23H,2,5-10H2,1,3-4H3/t17-,18?/m0/s1

InChI key

QHMBSVQNZZTUGM-ZENAZSQFSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.

Other Notes

EP issued certificate and SDS can be found here.
Sales restrictions may apply.


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library