Skip to Content
MilliporeSigma

SY3H7CF2AF22

Synthonix Corporation

(3aS,4S,5S,6R,7aS)-4-(ethoxycarbonyl)-6-ethyl-1-oxooctahydro-1H-indene-5-carboxylic acid

Synonym(s):

(3aS,4S,5S,6R,7aS)-4-(ethoxycarbonyl)-6-ethyl-1-oxooctahydro-1H-indene-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
282.34
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H22O5/c1-3-8-7-10-9(5-6-11(10)16)13(12(8)14(17)18)15(19)20-4-2/h8-10,12-13H,3-7H2,1-2H3,(H,17,18)/t8-,9+,10+,12+,13+/m1/s1

Inchi Key

JXRHNMLWDGTBKL-TZIOMAPQSA-N

Storage temp.

Refrigerator

Purity

95%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid



Still not finding the right product?


MSDS

https://synthonix.com/65868.html

pcodes

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library