Skip to Content
MilliporeSigma

TA9H11E41940

Targetmol Chemicals Inc.

16,17-Epoxypregnenol

Synonym(s):

1-[(2S,4aR,4bS,6aS,6bS,7aR,8aS,8bR)-2-hydroxy-4a,6a-dimethyl-1,2,3,4,4a,4b,5,6,6a,7a,8,8a,8b,9-tetradecahydro-6bH-naphtho[2',1':4,5]indeno[1,2-b]oxiren-6b-yl]ethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C21H30O3/c1-12(22)21-18(24-21)11-17-15-5-4-13-10-14(23)6-8-19(13,2)16(15)7-9-20(17,21)3/h4,14-18,23H,5-11H2,1-3H3/t14-,15+,16-,17-,18+,19-,20-,21+/m0/s1

Inchi Key

UQVIXFCYKBWZPJ-XXHSLLPRSA-N

Storage temp.

-80/-20

Purity

99.93%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid



Still not finding the right product?

Explore all of our products under 16,17-Epoxypregnenol


MSDS

https://www.targetmol.com/compound/16,17-Epoxypregnenol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library