Skip to Content
MilliporeSigma

TA9H93CFBE4D

Targetmol Chemicals Inc.

Ginsenoside C-K

Synonym(s):

(3beta,12beta)-3,12-dihydroxydammar-24-en-20-yl beta-D-glucopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C36H62O8/c1-20(2)10-9-14-36(8,44-31-30(42)29(41)28(40)23(19-37)43-31)21-11-16-35(7)27(21)22(38)18-25-33(5)15-13-26(39)32(3,4)24(33)12-17-34(25,35)6/h10,21-31,37-42H,9,11-19H2,1-8H3/t21-,22+,23+,24-,25+,26-,27-,28+,29-,30+,31-,33-,34+,35+,36-/m0/s1

Inchi Key

FVIZARNDLVOMSU-IRFFNABBSA-N

Storage temp.

-80/-20

Purity

99.97%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid



Still not finding the right product?

Explore all of our products under Ginsenoside C-K


MSDS

https://www.targetmol.com/compound/Ginsenoside C-K



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library