Skip to Content
MilliporeSigma

TA9H9A9A75C1

Targetmol Chemicals Inc.

Terpineol

Synonym(s):

1-methyl-4-(prop-1-en-2-yl)cyclohexan-1-ol--1-methyl-4-(propan-2-ylidene)cyclohexan-1-ol--2-(4-methylcyclohex-3-en-1-yl)propan-2-ol (1/1/1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/3C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;2*1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;11H,4-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

Inchi Key

NGVQETRGDCKEAH-UHFFFAOYSA-N

Storage temp.

-80/-20

Purity

98.15%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid



Still not finding the right product?

Explore all of our products under Terpineol


MSDS

https://www.targetmol.com/compound/Terpineol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library