Skip to Content
MilliporeSigma

BKCO-EG801-1

The Baker Company

Baker Company EdgeGARD e3 Horizontal Flow Clean Bench

8FT, Laminar Flow Clean Bench

Shipped & Sold By Baker Co

A Trusted Marketplace Seller

Synonym(s):

Laminar Flow Clean Bench, Horizontal Flow, Hood

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


manufacturer/tradename

The Baker Company

General description

DESCRIPTION

General Description

EdgeGARD® e3 8 ft Horizontal Flow Laminar Clean Bench
Ergonomically designed clean bench in 8-foot width. The EdgeGARD® e3 HF is a horizontal-flow clean bench that ensures product protection for a variety of life science and industrial laboratory and process applications. Directs HEPA-filtered airflow across the work bench to provide ISO Class 5 virtually particulate-free work area for preventing any contaminated room air from entering the work area.


Features:
  • 8 ft width
  • Standard Adjustable height stand allows for work surface height range of 27 1/2" to 41 3/8"
  • Work area depth of 20 5/16” and height of 34”
  • Interior workspace made from 16-gauge stainless steel
  • Satin finish work surface diminishes harsh light reflection
  • Slanted 10% façade allows operator more comfortable head and elbow position and reduces fatigue
  • Removable worksurface for increased sterility and easy cleaning
  • Front access to HEPA filters for quick and easy replacement
  • ISO Class 5 (Class 10) air cleanliness in the workspace
  • High-velocity return air slots maximize cleanliness and product protection
  • Washable pre-filter extends HEPA filter life


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library