MilliporeSigma
BKCO-REC602PLUS-174
The Baker Company
Baker Company ReCO2ver CO2 Incubator
Rapid recovery cell culture incubator with hydrogen peroxide biodecontamination and automatic CO2 Switch Over
Shipped & Sold By Baker Co
A Trusted Marketplace Seller
Synonym(s):
CO2 Incubator, Cell Culture Incubator, Lab incubator
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
The Baker Company
General description
DESCRIPTION
General Description
ReCO2ver™ rapid recovery cell culture Incubator with hydrogen peroxide biodecontamination and CO2 automatic supply switch over
The fastest and most precise recovery rates of cell culture conditions available. The ReCO2ver™ rapid recovery incubator provides the fastest recovery to set point times after door openings for CO2, humidity, and temperature. With active humidity control and a full-face HEPA filter, the ReCO2ver™ incubator is designed for mitigating contamination risk.
Features:
  • Control over temperature, carbon dioxide, & relative humidity
  • Full-faced HEPA filter
  • ISO Class 4 air cleanliness
  • CO2 Supply automatic switch over to prevent running out of gas
  • H2O2 biodecontamination in under 4 hour cycle
  • Exterior water reservoir and auto refill system – no need to open door to refill
  • UV to protect internal water pan from contamination
  • USB flash drive and alarm status relay
  • Interior workspace made from 16-gauge stainless steel
  • Work area depth of 20 5/16” and height of 34”
  • Fogless Interior Door
  • Uniform downward airflow
  • Active humidity control
  • 4 shelves standard
  • Crevice-free interior with coved corners for easy cleaning
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)