MilliporeSigma
NovaSeptum® GO Systems for Multi-use Bioprocess Sampling
Sampling manifold with multiple different containers connected to a stainless steel bioreactor via tubing and a blue and white holder. Containers include bags, a syringe, a bottle, and a tube with samples in orange-colored cell culture medium
NovaSeptum® GO sterile sampling systems are closed manual sampling solutions designed for multi-use or stainless steel processes. These innovative systems safeguard your sample integrity, minimize the risk of cross contamination from collection to analysis, and can be used for sampling throughout the drug manufacturing process.
Benefits of NovaSeptum® GO Sampling Systems
Quality you can trust: Leveraging our global quality systems and the Emprove® Program, our NovaSeptum® GO Sampling Systems come withextensive documentation and regulatory support, facilitating your risk assessment and process control.
Scientist in lab gear looking at a rack of tubes filled with different colored liquid
Representative Sampling
Locking tag and safety ring avoids accidental actuation and ensures process integrity and representative sampling.
Scientist with blue gloves is connecting two sampling tubes with a connector
Closed, Easy to Use and Validate
Ergonomically friendly and presterilized, the system improves operational efficiency and maintains sample integrity while reducing the possibility of sample loss.
A bioreactor as it is used in large scale pharmaceutical manufacturing with different sampling containers.
Flexible Sampling Configurations
Multiple containers of different size options in single and manifold designs that can be preconfigured or configured on site. 
If you are using MAST® Autosampling Solution, the NovaSeptum® sampling technologies are the preferred option for secure sample collection for sterility, bioburden or endotoxin testing.
Configure Your NovaSeptum® GO Sterile Sampling System
Step 1
Collection of industrial valves on a white background, featuring seven valves with metallic bodies and blue elements.
Select Your Holder
Choose the holder that fits your process: Tri-clamp (TC), In-line or Ingold®. TC holders are available preloaded with various containers.
Step 2
Assorted plastic containers of different shapes and volumes, some connected with tubing of different lengths and thicknesses. Orange and blue caps seal ends of tubing.
Choose Your Container Format
Choose from a large range of containers, sizes and configurations to meet all your sampling needs. Containers includes bottles, bags, syringes, conical tubes and transfer tubing.
Step 3
Metal tool on white background with a space at one end where tubing should be placed.  Tool designed for manual use with gloved hands.
Add a Crimper
Disconnect safely and maintain sterility using the Novaseptum® manual crimping tool.
Step 4
Multiple blue and white port plugs that are used as optional accessory with NovaSeptum® GO sampling units
Optional Accessories
Designed to maximize ease of use, and includes bag racks, port plugs, and locking tags.
Holder Information
While many configurations are available as standard, holders can be customized to particular sampling needs.
White plastic holder with blue plastic band and connection  points for NovaSeptum® GO sampling system. Plastic security tag prevents accidental actuation.
TC Holder
  • 1/2” to 2” size with up to 10 independent access points
  • Compatible with autoclave and steam in place sterilization
  • Available in single- or multi-use formats, preloaded with high purity bags, bottles or accurate volume syringes
Holder for NovaSeptum® GO system Ingold fitting with metal base and blue and white plastic components. Plastic locking indicator next to the holder.
Ingold® Holder
  • Available in any length
  • 2 independent points of access
  • Compatible with autoclave and steam in place sterilization
Stainless steel holder with blue plastic top for inline sample collection. Plastic locking indicator lying next to the holder.
In-line Holder
  • Ideal for collecting numerous samples between equipment
  • In-line 0.5” TC or butt-end holder available
  • Available for 0.5”, 0.75”, 13 mm or 19 mm tubing size
  • Compatible with autoclave and steam-in-place sterilization
Choose Your Container
Standard NovaSeptum® GO Sampling Units are single and manifold units of five containers available in bags, bottles, syringes, conical tubes or transfer units. Most NovaSeptum® GO standard assemblies are stocked and ready-to-ship. If the standard designs do not meet your requirements, connect with a Technical Specialist to design a custom solution tailored for your sampling need.
Find additional information on containers in our Technical Article "Sterile Sampling Containers for Representative Sampling and Reduced Risk".
Sterile sampling system with five manifolded plastic bags. Large bag at one end and tubing at other end.
High Purity Bag
  • Pureflex® bag. Polyethylene contact layer
  • 50 | 100 | 250 | 1000 mL
View Products
Sterile sampling system with five manifolded plastic bottles. Large drainage bag at one end and tubing at other end of unit.
Bottle
  • Polyethylene terephthalate glycol (PETG) bottle
  • 60 | 125 | 250 | 500 mL
View Products
Sterile sampling system with five manifolded syringes. Large drainage bag at one end and tubing at other end of unit.
Accurate Volume Syringe
  • Polycarbonate and Silicone
  • 5 | 20 mL
View Products
Sterile multi sampling system with five manifolded conical tubes. Large drainage bag at one end and tubing at other end of unit.
Conical Tube
  • 15 mL Polystyrene, 50 mL Polypropylene
  • Offered with or without dip tube
View Products
Sterile sampling transfer unit intended for transfer applications, with a needle on one end, tubing, and a tube outlet connection on the other end.
Transfer Units
  • Available in thermoplastic Elastomer | Silicone | C-Flex® tubing
View Products
Aseptic Disconnection
Metal tool on white background with a space at one end where tubing should be placed. Tool designed for manual use with gloved hands.
The NovaSeal® crimping tool securely seals and separates a pinch-piped tube in one simple motion. The pinch-pipe/tubing combination, along with the crimping tool, enables leak free disconnection of your single-use assembly and provides a safe way to separate your sample from the NovaSeptum® assembly without compromising sterility.
View Product
Optional Accessories
A metal and plastic bag rack with multiple metal hangers, intended to hold NovaSeptum® GO sampling units when loading or unloading.
Bag Rack
  • Easily load and unload NovaSeptum® sampling units.
  • For TC Holder, single or multi-sampling units.
View Products
Transparent plastic case fitted to hold a NovaSeptum® plastic bag when transporting, storing, or freezing.
Case
  • For transport, storage and freezing of
    50 | 100 | 250 mL High Purity Bag
View Products
Multiple blue and white port plugs that are used as optional accessory with NovaSeptum® GO sampling units
Other Accessories
  • Port Plug: For unused ports, easy to spot and minimizes risk of leakage
  • Volume Indicator: For High Purity Bags
  • Protector Caps: For crimped tubes
  • Centrifuge Caps: For 15 and 50 mL conical tubes
View Products